Assay Details
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031571.2 | NP_113759.1 | 6 - 7 | 841 | 103 | |
XM_008761703.1 | XP_008759925.1 | 7 - 8 | 920 | 103 | ||
GenBank mRNA | L10639.1 | - | 6 - 7 | 824 | 103 | |
S48190.1 | - | 6 - 7 | 841 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
