Assay Details
Gene Symbol: Tnfaip6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053382.1 | NP_445834.1 | 3 - 4 | 402 | 73 | |
GenBank mRNA | BC158666.1 | - | 3 - 4 | 402 | 73 | |
FQ228771.1 | - | 3 - 4 | 464 | 73 | ||
FQ228783.1 | - | 3 - 4 | 464 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
