Assay Details
Gene Symbol: Gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FM134560.1 | - | 3 - 4 | 433 | 86 | |
FM135509.1 | - | 4 - 5 | 562 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P236656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63311 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236656 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236656 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P236656 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P236656 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70252 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10939313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939315 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939316 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939312 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939311 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10939310 | - | - |
Rat Genome 230 2.0 Array | 1382063_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373199_at | - | - |
Rat Genome 230 2.0 Array | 1382063_at | - | - |
Rat Genome 230 2.0 Array | 1373199_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373199_at | - | - |
Affymetrix HT Rat Focus Array | 1373199_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382063_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382063_at | - | - |
Rat Expression Set 230 | 1373199_at | - | - |
Rat Expression Set 230 | 1373199_PM_at | - | - |
Rat Expression Set 230 | 1382063_PM_at | - | - |
Rat Expression Set 230 | 1382063_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939310 | - | - |
Rat Genome U34 Set | rc_AA943573_at | - | - |
Rat Genome U34 Set | rc_AI176183_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359658 | - | - |
