Assay Details
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235831.2 | XP_006235893.2 | 5 - 6 | 721 | 71 | |
XM_008762585.1 | XP_008760807.1 | 5 - 6 | 721 | 71 | ||
XM_008762586.1 | XP_008760808.1 | 5 - 6 | 721 | 71 | ||
XM_008762588.1 | XP_008760810.1 | 4 - 5 | 649 | 71 | ||
XM_008762589.1 | XP_008760811.1 | 5 - 6 | 721 | 71 | ||
XM_008762590.1 | XP_008760812.1 | 5 - 6 | 721 | 71 | ||
XM_008762591.1 | XP_008760813.1 | 5 - 6 | 721 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
