Assay Details
Gene Symbol: Eef1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012660.2 | NP_036792.2 | 1 - 2 | 250 | 124 | |
GenBank mRNA | BC074016.1 | - | 1 - 2 | 250 | 124 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC074016.1, NM_012660.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
