Assay Details
Gene Symbol: Dda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134790.1 | NP_001128262.1 | 2 - 3 | 191 | 80 | |
XM_006252948.2 | XP_006253010.1 | 2 - 3 | 1657 | 80 | ||
GenBank mRNA | BF549138.1 | - | 2 - 3 | 204 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
