Assay Details
Gene Symbol: Dda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134790.1 | NP_001128262.1 | 1 - 2 | 116 | 125 | |
GenBank mRNA | BF549138.1 | - | 1 - 2 | 129 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134790.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
