Assay Details
Gene Symbol: Rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107299.1 | NP_001100769.1 | 4 - 5 | 551 | 112 | |
XM_008771101.1 | XP_008769323.1 | 4 - 5 | 539 | 112 | ||
XM_008771102.1 | XP_008769324.1 | 4 - 5 | 512 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
