Assay Details
Gene Symbol: Rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107299.1 | NP_001100769.1 | 6 - 7 | 785 | 63 | |
XM_008771101.1 | XP_008769323.1 | 6 - 7 | 773 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
