Assay Details
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230081.2 | XP_006230143.1 | 2 - 3 | 751 | 77 | |
XM_006230082.2 | XP_006230144.1 | 2 - 3 | 751 | 77 | ||
XM_006230084.2 | XP_006230146.1 | 2 - 3 | 530 | 77 | ||
XM_008759769.1 | XP_008757991.1 | 2 - 3 | 751 | 77 | ||
GenBank mRNA | FQ219437.1 | - | 1 - 2 | 54 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
