Assay Details
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108499.1 | NP_001101969.1 | 1 - 2 | 232 | 112 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108499.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
