Assay Details
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019354.3 | NP_062227.2 | 6 - 7 | 1008 | 67 | |
GenBank mRNA | AB005143.1 | - | 4 - 5 | 635 | 67 | |
AB006613.1 | - | 4 - 5 | 635 | 67 | ||
AB010743.1 | - | 6 - 7 | 979 | 67 | ||
AF039033.1 | - | 4 - 5 | 635 | 67 | ||
BC062230.1 | - | 6 - 7 | 980 | 67 | ||
FQ227097.1 | - | 6 - 7 | 1009 | 67 | ||
FQ230356.1 | - | 2 - 3 | 294 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
