Assay Details
Gene Symbol: Nfatc2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007692.1 | NP_001007693.1 | 5 - 6 | 954 | 78 | |
GenBank mRNA | BC079050.1 | - | 5 - 6 | 954 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
