Assay Details
Gene Symbol: Bag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011936.1 | NP_001011936.1 | 2 - 3 | 615 | 71 | |
GenBank mRNA | BC085857.1 | - | 2 - 3 | 615 | 71 | |
DQ631552.1 | - | 2 - 3 | 618 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
