Assay Details
Gene Symbol: Stk33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759823.1 | XP_008758045.1 | 3 - 4 | 775 | 78 | |
XM_008759824.1 | XP_008758046.1 | 2 - 3 | 480 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
