Assay Details
Gene Symbol: Nsmce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039611.1 | NP_001034700.1 | 7 - 8 | 859 | 68 | |
XM_006230291.2 | XP_006230353.1 | 6 - 7 | 701 | 68 | ||
XM_006230292.2 | XP_006230354.1 | 6 - 7 | 1271 | 68 | ||
XM_006230293.2 | XP_006230355.1 | 6 - 7 | 781 | 68 | ||
GenBank mRNA | BC099757.1 | - | 7 - 8 | 859 | 5623851 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
