Assay Details
Gene Symbol: Cyb5r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014244.1 | NP_001014266.1 | 8 - 9 | 800 | 78 | |
XM_008759774.1 | XP_008757996.1 | 6 - 7 | 2050 | 78 | ||
XM_008759775.1 | XP_008757997.1 | 7 - 8 | 1953 | 78 | ||
XM_008759776.1 | XP_008757998.1 | 7 - 8 | 1965 | 78 | ||
XM_008759777.1 | XP_008757999.1 | 6 - 7 | 1938 | 78 | ||
XM_008759778.1 | XP_008758000.1 | 7 - 8 | 1368 | 78 | ||
XM_008759779.1 | XP_008758001.1 | 7 - 8 | 1525 | 78 | ||
XM_008759780.1 | XP_008758002.1 | 5 - 6 | 554 | 78 | ||
XM_008759781.1 | XP_008758003.1 | 8 - 9 | 849 | 78 | ||
XM_008759782.1 | XP_008758004.1 | 6 - 7 | 1986 | 78 | ||
GenBank mRNA | BC079235.1 | - | 8 - 9 | 800 | 7132617 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
