Assay Details
Gene Symbol: Lcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199405.2 | NP_955437.1 | 10 - 11 | 1088 | 92 | |
GenBank mRNA | BC063186.1 | - | 10 - 11 | 1091 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein methylation
C-terminal protein methylation
regulation of glucose metabolic process
negative regulation of protein-containing complex assembly
methylation
regulation of apoptotic process
regulation of mitotic cell cycle spindle assembly checkpoint
protein C-terminal carboxyl O-methyltransferase activity
methyltransferase activity
protein methyltransferase activity
protein methylation
C-terminal protein methylation
regulation of glucose metabolic process
negative regulation of protein-containing complex assembly
methylation
regulation of apoptotic process
regulation of mitotic cell cycle spindle assembly checkpoint
protein C-terminal carboxyl O-methyltransferase activity
methyltransferase activity
protein methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P233592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71891 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P233592 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P233592 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P233592 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1013959 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P233592 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1564 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7452 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35973 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5431 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63533 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10710741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710745 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710750 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710751 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710749 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710747 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10710739 | - | - |
Rat Genome 230 2.0 Array | 1388747_at | - | - |
Rat Genome 230 2.0 Array | 1388747_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388747_at | - | - |
Affymetrix HT Rat Focus Array | 1388747_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100019747.rn.1 | TC0100002544.rn.1 | RTA TC |
Rat Expression Set 230 | 1388747_at | - | - |
Rat Expression Set 230 | 1388747_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710739 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21786301 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366574 | - | - |
RatRef-12 Expression BeadChip | ILMN_51502 | - | - |
