Assay Details
Gene Symbol: N4bp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035222.2 | NP_001030299.1 | 3 - 4 | 372 | 59 | |
XM_008768992.1 | XP_008767214.1 | 3 - 4 | 372 | 59 | ||
GenBank mRNA | BC168658.1 | - | 3 - 4 | 372 | 7314496 | 59 |
FQ231381.1 | - | 3 - 4 | 343 | 59 | ||
FQ231652.1 | - | 3 - 4 | 343 | 59 | ||
FQ235206.1 | - | 3 - 4 | 343 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
