Assay Details
Gene Symbol: Mastl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107369.1 | NP_001100839.1 | 2497 | 99 | ||
XM_006254340.2 | XP_006254402.1 | 2562 | 99 | |||
XM_006254341.2 | XP_006254403.1 | 2424 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
mitotic cell cycle
protein phosphorylation
female meiosis II
biological_process
negative regulation of phosphoprotein phosphatase activity
regulation of cell cycle
molecular_function
protein serine/threonine kinase activity
ATP binding
kinase activity
G2/M transition of mitotic cell cycle
mitotic cell cycle
protein phosphorylation
female meiosis II
biological_process
negative regulation of phosphoprotein phosphatase activity
regulation of cell cycle
molecular_function
protein serine/threonine kinase activity
ATP binding
kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P213149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52216 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P213149 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P213149 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P213149 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P213149 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86575 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9115 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10800004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800010 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800005 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800000 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800002 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10799999 | - | - |
Affymetrix Rat Clariom S Assay | PSR1700007907.rn.1 | TC1700001142.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700007906.rn.1 | TC1700001142.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10799999 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20753157 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359099 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373768 | - | - |
