Assay Details
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106580.2 | NP_001100050.1 | 2 - 3 | 452 | 87 | |
XM_006236029.2 | XP_006236091.1 | 2 - 3 | 505 | 87 | ||
XM_006236030.2 | XP_006236092.1 | 2 - 3 | 534 | 87 | ||
GenBank mRNA | BC168741.1 | - | 2 - 3 | 452 | 6919194 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
