Assay Details
Gene Symbol: Hepacam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106580.2 | NP_001100050.1 | 4 - 5 | 1034 | 70 | |
XM_006236029.2 | XP_006236091.1 | 4 - 5 | 1087 | 70 | ||
GenBank mRNA | BC168741.1 | - | 4 - 5 | 1034 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
