Assay Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232168.1 | XP_006232230.1 | 3 - 4 | 455 | 117 | |
XM_008760873.1 | XP_008759095.1 | 3 - 4 | 455 | 117 | ||
XM_008760874.1 | XP_008759096.1 | 2 - 3 | 3086 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
