Assay Details
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013910.2 | NP_001013932.1 | 3 - 4 | 505 | 153 | |
GenBank mRNA | AY552804.1 | - | 3 - 4 | 432 | 153 | |
BC097370.1 | - | 3 - 4 | 505 | 7444022 | 153 | |
DQ120489.1 | - | 3 - 4 | 397 | 153 | ||
DQ120490.1 | - | 3 - 4 | 397 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
