Assay Details
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013910.2 | NP_001013932.1 | 4 - 5 | 575 | 81 | |
GenBank mRNA | AY552804.1 | - | 4 - 5 | 502 | 81 | |
BC097370.1 | - | 4 - 5 | 575 | 7444022 | 81 | |
DQ120489.1 | - | 4 - 5 | 467 | 81 | ||
DQ120490.1 | - | 4 - 5 | 467 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
