Assay Details
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013910.2 | NP_001013932.1 | 5 - 6 | 770 | 66 | |
GenBank mRNA | AY552804.1 | - | 5 - 6 | 697 | 66 | |
BC097370.1 | - | 5 - 6 | 770 | 66 | ||
DQ120489.1 | - | 5 - 6 | 662 | 66 | ||
DQ120490.1 | - | 5 - 6 | 662 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
