Assay Details
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760849.1 | XP_008759071.1 | 3 - 4 | 1596 | 129 | |
XM_574892.6 | XP_574892.1 | 4 - 5 | 1067 | 129 | ||
GenBank mRNA | U31668.1 | - | 4 - 5 | 416 | 129 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
