Assay Details
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760849.1 | XP_008759071.1 | 5 - 6 | 1922 | 84 | |
XM_574892.6 | XP_574892.1 | 6 - 7 | 1393 | 84 | ||
GenBank mRNA | U31668.1 | - | 6 - 7 | 742 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
