Assay Details
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760849.1 | XP_008759071.1 | 6 - 7 | 1977 | 104 | |
XM_574892.6 | XP_574892.1 | 7 - 8 | 1448 | 104 | ||
GenBank mRNA | U31668.1 | - | 7 - 8 | 797 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
