Assay Details
Gene Symbol: Mrpl47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037183.1 | NP_001032260.1 | 5 - 6 | 543 | 65 | |
XM_008760912.1 | XP_008759134.1 | 4 - 5 | 380 | 65 | ||
GenBank mRNA | BC089884.1 | - | 5 - 6 | 544 | 65 | |
BC105820.1 | - | 5 - 6 | 543 | 7369885 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
