Assay Details
Gene Symbol: Dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107668.1 | NP_001101138.1 | 1 - 2 | 146 | 101 | |
XM_006232262.2 | XP_006232324.1 | 1 - 2 | 357 | 101 |
Gene Symbol: LOC102553014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760957.1 | XP_008759179.1 | 1 - 2 | 428 | 101 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107668.1,XM_006232262.2,XM_008760957.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
