Assay Details
Gene Symbol: Dcun1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107668.1 | NP_001101138.1 | 2 - 3 | 368 | 107 | |
XM_003749267.3 | XP_003749315.1 | 247 | 107 | |||
XM_006232261.2 | XP_006232323.1 | 3 - 4 | 366 | 107 | ||
XM_006232262.2 | XP_006232324.1 | 2 - 3 | 579 | 107 | ||
XM_006232263.2 | XP_006232325.1 | 2 - 3 | 294 | 107 | ||
XM_006232264.2 | XP_006232326.1 | 2 - 3 | 1034 | 107 | ||
XM_008760925.1 | XP_008759147.1 | 2 - 3 | 353 | 107 | ||
XM_008760926.1 | XP_008759148.1 | 385 | 107 | |||
XM_008760927.1 | XP_008759149.1 | 2 - 3 | 1806 | 107 |
Gene Symbol: LOC102553014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760957.1 | XP_008759179.1 | 2 - 3 | 650 | 107 | |
XM_008760958.1 | XP_008759180.1 | 2 - 3 | 245 | 107 | ||
XM_008760959.1 | XP_008759181.1 | 2 - 3 | 1034 | 107 | ||
XM_008760960.1 | XP_008759182.1 | 3 - 4 | 385 | 107 | ||
XM_008760961.1 | XP_008759183.1 | 3 - 4 | 370 | 107 | ||
XM_008760962.1 | XP_008759184.1 | 2 - 3 | 1808 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
