Assay Details
Gene Symbol: Ctif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108891.1 | NP_001102361.1 | 4 - 5 | 977 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P304727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303351 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48147 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102098 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P304727 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P304727 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P304727 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P304727 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32114 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37574 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10805244 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10805247 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10805246 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10805245 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10805243 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10805242 | - | - |
Affymetrix Rat Clariom S Assay | PSR1800011965.rn.1 | TC1800001711.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10805242 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21072009 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372816 | - | - |
