Assay Details
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127636.1 | NP_001121108.1 | 3 - 4 | 304 | 88 | |
XM_008770571.1 | XP_008768793.1 | 2 - 3 | 251 | 88 | ||
GenBank mRNA | BC166528.1 | - | 3 - 4 | 304 | 7189304 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008770571.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
