Assay Details
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127636.1 | NP_001121108.1 | 5 - 6 | 551 | 71 | |
XM_008770571.1 | XP_008768793.1 | 4 - 5 | 498 | 71 | ||
GenBank mRNA | BC166528.1 | - | 5 - 6 | 551 | 7189304 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
