Assay Details
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127636.1 | NP_001121108.1 | 7 - 8 | 781 | 89 | |
XM_008770571.1 | XP_008768793.1 | 6 - 7 | 728 | 89 | ||
GenBank mRNA | BC166528.1 | - | 7 - 8 | 781 | 7189304 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
