Assay Details
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127636.1 | NP_001121108.1 | 1 - 2 | 68 | 88 | |
GenBank mRNA | BC166528.1 | - | 1 - 2 | 68 | 88 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166528.1, NM_001127636.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
