Assay Details
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109097.1 | NP_001102567.1 | 1 - 2 | 64 | 61 | |
GenBank mRNA | FQ214748.1 | - | 1 - 2 | 64 | 61 | |
FQ214808.1 | - | 1 - 2 | 64 | 61 | ||
FQ214853.1 | - | 1 - 2 | 47 | 61 | ||
FQ215077.1 | - | 66 | 61 | |||
FQ215190.1 | - | 1 - 2 | 66 | 61 | ||
FQ215382.1 | - | 1 - 2 | 66 | 61 | ||
FQ215728.1 | - | 1 - 2 | 66 | 61 | ||
FQ215901.1 | - | 1 - 2 | 68 | 61 | ||
FQ215918.1 | - | 1 - 2 | 64 | 61 | ||
FQ216019.1 | - | 1 - 2 | 104 | 61 | ||
FQ216101.1 | - | 1 - 2 | 66 | 61 | ||
FQ216115.1 | - | 1 - 2 | 65 | 61 | ||
FQ216248.1 | - | 1 - 2 | 65 | 61 | ||
FQ216396.1 | - | 1 - 2 | 48 | 61 | ||
FQ216753.1 | - | 1 - 2 | 48 | 61 | ||
FQ216842.1 | - | 1 - 2 | 48 | 61 | ||
FQ224894.1 | - | 1 - 2 | 68 | 61 | ||
FQ224977.1 | - | 1 - 2 | 64 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109097.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
