Assay Details
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109097.1 | NP_001102567.1 | 2 - 3 | 158 | 82 | |
XM_006251655.2 | XP_006251717.1 | 2 - 3 | 205 | 82 | ||
GenBank mRNA | FQ214748.1 | - | 2 - 3 | 158 | 82 | |
FQ214808.1 | - | 2 - 3 | 158 | 82 | ||
FQ214853.1 | - | 2 - 3 | 141 | 82 | ||
FQ215077.1 | - | 160 | 82 | |||
FQ215190.1 | - | 2 - 3 | 160 | 82 | ||
FQ215360.1 | - | 2 - 3 | 164 | 82 | ||
FQ215382.1 | - | 2 - 3 | 160 | 82 | ||
FQ215728.1 | - | 2 - 3 | 160 | 82 | ||
FQ215901.1 | - | 2 - 3 | 162 | 82 | ||
FQ215918.1 | - | 2 - 3 | 158 | 82 | ||
FQ216019.1 | - | 2 - 3 | 198 | 82 | ||
FQ216101.1 | - | 2 - 3 | 160 | 82 | ||
FQ216115.1 | - | 2 - 3 | 159 | 82 | ||
FQ216248.1 | - | 2 - 3 | 159 | 82 | ||
FQ216396.1 | - | 2 - 3 | 142 | 82 | ||
FQ216753.1 | - | 2 - 3 | 142 | 82 | ||
FQ217380.1 | - | 180 | 82 | |||
FQ224894.1 | - | 2 - 3 | 162 | 82 | ||
FQ224977.1 | - | 2 - 3 | 158 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
