Assay Details
Gene Symbol: Abhd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007680.1 | NP_001007681.1 | 2 - 3 | 195 | 71 | |
XM_006251766.2 | XP_006251828.1 | 2 - 3 | 1474 | 71 | ||
XM_006251767.2 | XP_006251829.1 | 2 - 3 | 1006 | 71 | ||
XM_006251768.2 | XP_006251830.1 | 2 - 3 | 171 | 71 | ||
GenBank mRNA | BC083826.1 | - | 2 - 3 | 195 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
