Assay Details
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240772.2 | XP_006240834.1 | 4 - 5 | 583 | 64 | |
XM_006240773.2 | XP_006240835.1 | 4 - 5 | 645 | 64 | ||
XM_006240774.2 | XP_006240836.1 | 3 - 4 | 491 | 64 | ||
XM_006240775.2 | XP_006240837.1 | 4 - 5 | 501 | 64 | ||
XM_006240776.2 | XP_006240838.1 | 3 - 4 | 409 | 64 | ||
XM_008765030.1 | XP_008763252.1 | 3 - 4 | 1502 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
