Assay Details
Gene Symbol: Prkar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033679.1 | NP_001028851.2 | 8 - 9 | 889 | 131 | |
XM_008768942.1 | XP_008767164.1 | 7 - 8 | 1162 | 131 | ||
GenBank mRNA | BC091371.1 | - | 3 - 4 | 408 | 131 | |
BC104679.1 | - | 8 - 9 | 889 | 131 | ||
FQ212815.1 | - | 7 - 8 | 997 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
