Assay Details
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108059.3 | NP_001101529.1 | 3 - 4 | 418 | 82 | |
XM_006256614.2 | XP_006256676.1 | 2 - 3 | 1400 | 82 | ||
GenBank mRNA | EF429098.1 | - | 1 - 2 | 216 | 82 | |
FM043339.1 | - | 3 - 4 | 424 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
