Assay Details
Gene Symbol: Angpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134454.1 | NP_604449.1 | 2 - 3 | 519 | 68 | |
GenBank mRNA | AY052400.1 | - | 1 - 2 | 82 | 68 | |
BC161931.1 | - | 2 - 3 | 519 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
