Assay Details
Gene Symbol: Angpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134454.1 | NP_604449.1 | 4 - 5 | 868 | 67 | |
GenBank mRNA | BC161931.1 | - | 4 - 5 | 868 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
