Assay Details
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134710.1 | NP_001128182.1 | 3 - 4 | 388 | 65 | |
GenBank mRNA | BC166762.1 | - | 3 - 4 | 388 | 7931606 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
