Assay Details
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008859.1 | NP_001008859.1 | 3 - 4 | 228 | 90 | |
XM_006244464.2 | XP_006244526.1 | 4 - 5 | 377 | 90 | ||
XM_006244465.2 | XP_006244527.1 | 4 - 5 | 374 | 90 | ||
GenBank mRNA | AY310148.1 | - | 3 - 4 | 228 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
