Assay Details
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244464.2 | XP_006244526.1 | 1 - 2 | 100 | 88 | |
XM_006244465.2 | XP_006244527.1 | 1 - 2 | 100 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
