Assay Details
Gene Symbol: Supt3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766907.1 | XP_008765129.1 | 1 - 2 | 235 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766907.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
