Assay Details
Gene Symbol: Supt3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244587.2 | XP_006244649.2 | 9 - 10 | 1571 | 81 | |
XM_008766906.1 | XP_008765128.1 | 9 - 10 | 1571 | 81 | ||
XM_008766907.1 | XP_008765129.1 | 9 - 10 | 1044 | 81 | ||
XM_008766908.1 | XP_008765130.1 | 9 - 10 | 940 | 81 | ||
XM_008766909.1 | XP_008765131.1 | 9 - 10 | 901 | 81 | ||
XM_008766910.1 | XP_008765132.1 | 8 - 9 | 814 | 81 | ||
XM_008766911.1 | XP_008765133.1 | 8 - 9 | 799 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
